Order Entry
ContactUsLinkComponent
RED TAQ 2X MASTER MIX 1 5MM MG 500 R
VWR® RED Taq® DNA Polymerase Master Mix
# de catalogue 76620-472
Fournisseur:  VWR International
Numéro CAS:  
RED TAQ 2X MASTER MIX 1 5MM MG 500 R
VWR® RED Taq® DNA Polymerase Master Mix
# de catalogue 76620-472
Fournisseur:  VWR International
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Concentration:
    2X
  • Description:
    Red Taq DNA Polymerase 2X Master Mix (1.5 mM MgCl₂)
  • Units:
    Each
  • No. of Reactions:
    500 (of 50 μl)

Spécifications

A propos de cet article

VWR® Red Taq DNA Polymerase Master Mix, which also contains an inert red dye, can be directly loaded onto an agarose gel without addition of electrophoresis loading buffers.

  • All-in-one master mix for great convenience
  • Timesaving reaction setup
  • Direct loading onto agarose gel
  • Easy visualisation of pipetting
  • Red tracking dye
  • Increased reproducibility
  • High product yield
  • Processes up to 5 kb

Red Taq DNA Polymerase master mixes are a good alternative to VWR® Taq DNA Polymerase. The inert red dye and stabilizer are extra features, which allow direct loading onto your agarose gel for analysis.

The Red Taq DNA Polymerase master mixes offer reduced set-up time and eliminate the risk of contamination of reaction components. Fewer handling steps lead to better reproducibility, which makes master mixes suitable for standard PCR applications, screening and high-throughput testing.

Red Taq DNA Polymerase Master Mix is a ready-to-use 1.1X or 2X reaction mix. Simply add primers, template, and water to carry out primer extensions and other molecular biology applications.

Long term storage at -20 °C. Product expiry at -20 °C is stated on the label. Optional: Store at +4 °C for up to 6 months.

VWR® Red Taq DNA Polymerase 1.1X and 2X master mixes are available in two final MgCl2 concentrations: 1.5 mM and 2 mM.

Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought