Smiths Medical® Fitting, Polycarbonate, Three Ports, Manifolds with Female Luer Locks, 360° Rotation, Non-Sterile; 50/PK
# de catalogue MFLX30600-41BLK
Fournisseur: VWR International
Spécifications
Voir toutes les configurations de produits- Wetted materials:
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Warranty:
- Out of box
- Out of box
- Out of box
- En cours de visualisation
Spécifications
A propos de cet article
Regulate flow without any leaking.
- Three ports with female luer locks
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Polycarbonate manifolds have polyurethane stems and HDPE handles. Ports have female luer locks.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
- Description:Fitting, Polycarbonate, Three Ports, Manifolds with Female Luer Locks, 360° Rotation, Non-Sterile; 50/PK
- Wetted materials:Polycarbonate
- Warranty:Out of box
- Cat. no.:MFLX30600-41BLK
- Flow Pattern:3 Ports, 360 Degree Rotation
- Inlet Port:Female luer
- Outlet Port:Female luer
Customers Who Bought This Also Bought