Order Entry
ContactUsLinkComponent
NATIVE BARCODING AUCILIARY KIT V14
Native barcoding auxiliary kit v14, Oxford Nanopore Technologies
# de catalogue 76595-412
Numéro CAS:  
NATIVE BARCODING AUCILIARY KIT V14
Native barcoding auxiliary kit v14, Oxford Nanopore Technologies
# de catalogue 76595-412
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Compatible with:
    R10.4.1 flow cells and kit 14 chemistry
  • Description:
    Native barcoding auxiliary kit v14
  • Cat. no.:
    76595-412
  • No. of Reactions:
    12

Spécifications

A propos de cet article

This expansion is recommended for users who want to make use of unused barcodes to maximise the use of their Native Barcoding Kits.

This expansion pack contains additional library preparation reagents for the Native Barcoding Kit 24 and 96 V14 (SQK-NBD114.24 and SQK-NBD114.96) for another 12 reactions in conjunction with the Sequencing Auxiliary Vials V14 (EXP-AUX003).

The Native Barcoding Auxiliary Kit V14 (EXP-NBA114) can only be used with Kit 14 chemistry. This kit is not compatible with previous kits chemistries.

Ordering information: Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.
Caution: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.
Customers Who Bought This Also Bought