Spécifications
- Length:19.1 mm (0.75")
- Description:With 12" tubing
- Gauge:19
Spécifications
A propos de cet article
The Sol-Vet Butterfly Needle features a removeable multi-sample luer adapter, making its application flexible across multiple uses. This is a safety-engineered device with a sliding safety shield that clicks when activated, telling you the needle is safely locked.
- Thin-walled needle minimizes patient discomfort
- Transparent design allows for clear visualization of blood flow
- Available with or without pre-attached blood collection tube holder
Products for veterinary use only.