Spécifications
- Length:³/₄"
- Cat. no.:76582-494
- Gauge:24
Spécifications
A propos de cet article
The Sol-Vet I.V. Catheter is used for continuous and intermittent intravenous medication delivery. It is designed with a safety feature that covers the needle bevel, helping to prevent needlestick injuries.
- Protective casing ensures safety of catheter during transportation
- Translucent catheter hub and chamber allows for easy visualization of blood flashback
- Contains a hydrophobic membrane filter to prevent blood leakage during insertion
- Filter cap can be removed to allow connection with a syringe to assist insertion
Products for Veterinary use only.