ContactUsLinkComponent
CATHETER IV 24G 3/4IN BX100
Sol-Vet® IV Catheters (Veterinary Care), Sol M
# de catalogue 76582-494
Fournisseur:  SOL-MILLENNIUM MEDICAL
Sol-Vet® IV Catheters (Veterinary Care), Sol M
# de catalogue 76582-494
Fournisseur:  SOL-MILLENNIUM MEDICAL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Length:
    ³/₄"
  • Cat. no.:
    76582-494
  • Gauge:
    24
Spécifications
A propos de cet article
The Sol-Vet I.V. Catheter is used for continuous and intermittent intravenous medication delivery. It is designed with a safety feature that covers the needle bevel, helping to prevent needlestick injuries.
  • Protective casing ensures safety of catheter during transportation
  • Translucent catheter hub and chamber allows for easy visualization of blood flashback
  • Contains a hydrophobic membrane filter to prevent blood leakage during insertion
  • Filter cap can be removed to allow connection with a syringe to assist insertion
Products for Veterinary use only.
Customers Who Bought This Also Bought