ContactUsLinkComponent
SWABS ALCOHOL STERILE LRG BX100
Sol-M® Blood Collection Alcohol Swabs
# de catalogue 76582-864
Fournisseur:  SOL-MILLENNIUM MEDICAL
Sol-M® Blood Collection Alcohol Swabs
# de catalogue 76582-864
Fournisseur:  SOL-MILLENNIUM MEDICAL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Alcohol swabs, large (sterile)
  • Cat. no.:
    76582-864
Spécifications
A propos de cet article
70% isopropyl alcohol to clean and prepare the skin. Individually wrapped in 4-layer wrappers for an air-tight seal that prevents leakage and drying.
Customers Who Bought This Also Bought