ContactUsLinkComponent
HOLDER BLD COL TB W/VACUUM ACT BX50
Sol Care® Vacuum Activated Blood Collection Safety Tube Holder Set, Sol M
# de catalogue 76582-744
Fournisseur:  SOL-MILLENNIUM MEDICAL
HOLDER BLD COL TB W/VACUUM ACT BX50
Sol Care® Vacuum Activated Blood Collection Safety Tube Holder Set, Sol M
# de catalogue 76582-744
Fournisseur:  SOL-MILLENNIUM MEDICAL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Vacuum activated blood collection safety tube holder set
  • Cat. no.:
    76582-744
Spécifications
A propos de cet article
The Sol-Care Vacuum Activated Blood Collection Safety Tube Holder Set is an innovative, exclusive blood collection safety accessory.
  • Dominant hand remains on last tube and behind the proximal end of tube holder
  • Safety mechanism is integral part of device and doesn't block sight of needle tip
  • Hands remain behind needle during the point of greatest potential hazard
  • Complies with OSHA regulations to prevent cross-contamination
The unique retraction mechanism is built into the special tube. After all specimens are drawn, the tube is pushed into the needle/hub assembly and withdraws the used needle, protecting the healthcare worker from any chance of accidental needlestick. This new safety device requires no additional movements or changes in technique. The activation is accomplished using the same forward pressure as is customary when using all other blood collection tubes.
Customers Who Bought This Also Bought