Spécifications
- Description:Vacuum activated blood collection safety tube holder set
- Cat. no.:76582-744
Spécifications
A propos de cet article
The Sol-Care Vacuum Activated Blood Collection Safety Tube Holder Set is an innovative, exclusive blood collection safety accessory.
- Dominant hand remains on last tube and behind the proximal end of tube holder
- Safety mechanism is integral part of device and doesn't block sight of needle tip
- Hands remain behind needle during the point of greatest potential hazard
- Complies with OSHA regulations to prevent cross-contamination
The unique retraction mechanism is built into the special tube. After all specimens are drawn, the tube is pushed into the needle/hub assembly and withdraws the used needle, protecting the healthcare worker from any chance of accidental needlestick. This new safety device requires no additional movements or changes in technique. The activation is accomplished using the same forward pressure as is customary when using all other blood collection tubes.