ContactUsLinkComponent
SYRINGE LL 10ML W/O NEEDLE BX100
Sol-M® Standard Syringes without Needle
# de catalogue 76582-806
Fournisseur:  SOL-MILLENNIUM MEDICAL
Sol-M® Standard Syringes without Needle
# de catalogue 76582-806
Fournisseur:  SOL-MILLENNIUM MEDICAL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Sterility:
    Sterile
  • Volume:
    10 ml
  • Description:
    Syringe
  • Cat. no.:
    76582-806
Spécifications
A propos de cet article
3-Piece syringes accurately measure liquid medications and are available in a range of sizes and connection combinations to address a wide variety of clinical needs.
  • High-transparency barrel to easily visualize contents
  • Silicone lubrication ensures smooth plunger movement
  • Plunger backstop that prevents accidental withdrawal of plunger from the barrel
  • Large finger grips allow for easy, controlled injections
Available in Luer lock, Luer slip tip and catheter tip.
Customers Who Bought This Also Bought