Spécifications
- Sterility:Sterile
- Volume:10 ml
- Description:Syringe
- Cat. no.:76582-806
Spécifications
A propos de cet article
3-Piece syringes accurately measure liquid medications and are available in a range of sizes and connection combinations to address a wide variety of clinical needs.
- High-transparency barrel to easily visualize contents
- Silicone lubrication ensures smooth plunger movement
- Plunger backstop that prevents accidental withdrawal of plunger from the barrel
- Large finger grips allow for easy, controlled injections
Available in Luer lock, Luer slip tip and catheter tip.