VWR® Autoclavable Cleanroom Goggles
# de catalogue 76579-452
Fournisseur: VWR International
Spécifications
- Lens Coating:Anti-fog, anti-scratch
- Description:Cleanroom goggles
- Frame Color:White
Spécifications
A propos de cet article
Cleanroom goggles feature a white frame with indirect ventilation.
- Anti-fog and anti-scratch protection
- Clear lens
- Indirect ventilation
- Silicone strap
- White frame
- Autoclavable
- Tested to withstand >40 autoclave cycles (30 minutes duration at 121/250 °C)
Clear replacement lenses also feature anti-fog and anti-scratch protection.
Ordering information: Replacement lenses are sold separately.
Customers Who Bought This Also Bought