ContactUsLinkComponent
GOGGLES AUTOCLAVABLE PK10
VWR® Autoclavable Cleanroom Goggles
# de catalogue 76579-452
Fournisseur:  VWR International
GOGGLES AUTOCLAVABLE PK10
VWR® Autoclavable Cleanroom Goggles
# de catalogue 76579-452
Fournisseur:  VWR International
Spécifications
  • Lens Coating:
    Anti-fog, anti-scratch
  • Description:
    Cleanroom goggles
  • Frame Color:
    White
Spécifications
A propos de cet article
Cleanroom goggles feature a white frame with indirect ventilation.
  • Anti-fog and anti-scratch protection
  • Clear lens
  • Indirect ventilation
  • Silicone strap
  • White frame
  • Autoclavable
  • Tested to withstand >40 autoclave cycles (30 minutes duration at 121/250 °C)
Clear replacement lenses also feature anti-fog and anti-scratch protection.
Ordering information: Replacement lenses are sold separately.
Customers Who Bought This Also Bought