Ceramic Magnetic Hotplate Stirrer, HSC, VELP Scientifica
Catalog # 76559-352
Supplier: VELP SCIENTIFIC INC.
Specifications
- Temperature range:Ambient...400 °C
- Description:Hotplate stirrer analog HSC
- Model:HSC
- Max. stirring capacity H₂O:15 L
- Plate material:Ceramic
- Plate dimensions:180×180 mm (7×7")
- Speed range:50 - 1300 rpm
- Electrical:115 V / 60 Hz
- W×D×H:203×344×94 mm (8.0×13.5×3.7")
- Weight:3.2 kg (7.0 lbs.)
- Cat. no.:76559-352
Specifications
About this item
The entry level hotplate stirrer for general laboratory applications. You can rely on the HSC for everyday laboratory applications like sample and buffer solutions preparation, as it stirs volumes up to 15 liters (H₂O). HSC is equipped with a cutting-edge Heating Plate technology engineered by VELP that ensures temperature homogeneity, thermal efficiency and durable heating performance over time.
- Analog hotplate stirrer with ceramic plate and techno polymer housing
- Features ensure a high level of safety and protection for researchers and laboratory operators
- HSC stirs aqueous solutions from 50 up to 1300 rpm, an ideal solution for general laboratory applications
- The extremely resistant ceramic plate is easy to clean and resistant to acids, bases and solvents
- Monophase PCM motor
Customers Who Bought This Also Bought