CAP SCR 9 MM BL PP S/T BND PK100
# de catalogue TSCHSC9-35UB
Fournisseur: Thermo Scientific
Customers Who Bought This Also Bought
Potassium dihydrogen phosphate ≥99.0%, GR ACS, Supelco®
Fournisseur: MilliporeSigma
$1,033.55 to $4,052.62
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$194.14 to $694.12