SureSTART™ Manual Crimper for 20 mm Crimp Vials, Level 3, Thermo Scientific
# de catalogue TS60180-CR11
Fournisseur: Thermo Scientific
Spécifications
- Description:Manual crimper, red, suitable for 11 mm crimp caps
- Cat. no.:TS60180-CR11
Spécifications
A propos de cet article
Use SureSTART™ level 3 manual vial crimpers for 20 mm Crimp vials to ensure that your crimp caps are sealed reliably every time.
Customers Who Bought This Also Bought