SureSTART™ Septa for 20 mm Stoppers, Level 2 High-throughput Applications, Thermo Scientific
# de catalogue TS6A20ST1T
Fournisseur: Thermo Scientific
Spécifications
- Ø:20 mm
- Septum:White silicone/beige PTFE
- Description:Septum
- Thickness:3.0 mm
- Cat. no.:TS6A20ST1T
- Packaging:10×100
Spécifications
A propos de cet article
Use Thermo Scientific™ SureSTART™ 20 mm stoppers with crimp vials that have a 20 mm opening and a secure push fit, including our SureSTART 20 mm crimp headspace vials (level 2).
Customers Who Bought This Also Bought