Spoon Spatulas, 9"
# de catalogue 470346-060
Fournisseur: Wards
Spécifications
- Description:9" spoon spatulas, 6/pack
- Cat. no.:470346-060
Spécifications
A propos de cet article
Polished stainless steel spatulas with a wide end (³/₄") and narrow end (⁵/₁₆"), individually wrapped for hygiene, making them perfect for laboratory use or culinary applications.
- Spoon and spatula ends
- High quality, individually wrapped
Spoon-spatula tools (6). These spoon spatula tools are high quality and perfect for a variety of applications.
Customers Who Bought This Also Bought