Order Entry
ContactUsLinkComponent
CARBOY BOTTLE WITH STOPCOCK 1
Eisco Polypropylene Carboy Bottle with Stopcock
# de catalogue 470344-684
Fournisseur:  Wards
CARBOY BOTTLE WITH STOPCOCK 1
Eisco Polypropylene Carboy Bottle with Stopcock
# de catalogue 470344-684
Fournisseur:  Wards

Spécifications

  • Capacity:
    10000 ml
  • O.D.:
    10"
  • Height:
    15"
  • Cat. no.:
    470344-684

Spécifications

A propos de cet article

Large carboy bottles for convenient dispensing of liquids.

  • Made of heat tolerant and chemical resistant polypropylene plastic
  • Leak-proof stopcock spigot with gasket screwed into the container to regulated liquid flow
  • Features two integral molded handles for easy transport

Ideal for laboratory use. Features a leak-proof stopcock spigot with gasket screwed into the container to regulated liquid flow and two handles for easy maneuverability and transport.

Customers Who Bought This Also Bought