ContactUsLinkComponent
WASH BOTTLE WATER
Wash Bottle for Water
# de catalogue 470344-058
Fournisseur:  Wards
WASH BOTTLE WATER
Wash Bottle for Water
# de catalogue 470344-058
Fournisseur:  Wards
Spécifications
  • Capacity:
    500 ml
  • Description:
    Wash bottle for water
  • Cat. no.:
    470344-058
Spécifications
A propos de cet article
Wash bottle for water with color-coded labels, wide mouth, self-venting design. Bottle is made of durable polypropylene, ideal for laboratory use with easy and precise dispensing.
  • Easy to read labeling, including chemical information and precautionary statements
  • Blue cap with built-in nozzle
  • Wide mouth for easy filling
  • Self venting design to eliminate leaks and dripping
  • Easy to control squeeze for dispensing
Made of low density polyethylene (LDPE) for superior quality and strength. Innovative flex ink won't fade, crack, or peel. Bottle is labeled for water and features chemical information, as well as safe handling and precautionary statements. Wide mouth for easy refilling. Each bottle features a two-piece construction, consisting of the bottle and leak-proof cap with built-in nozzle. Self venting design eliminates leaks and dripping. The low density polyethylene plastic is very responsive, with smooth bounce back that won't get stiff over time.
Each bottle measures 9" in total height (5.25" bottle alone) and 2.9" in diameter. Bottle opening has an inner diameter of 1.7" and an outer diameter measuring 1.9".
Customers Who Bought This Also Bought