ContactUsLinkComponent
SW VWR CHASE WORKSU MOUNT 42H 6W
VWR® Atlas Pipe Chase Assemblies
# de catalogue VWRPC426-W
Fournisseur:  VWR International
VWR® Atlas Pipe Chase Assemblies
# de catalogue VWRPC426-W
Fournisseur:  VWR International
Spécifications
  • Color:
    Snow white
  • Description:
    Counter-Mounted Pipe Chase Assembly
  • Cat. no.:
    VWRPC426-W
Spécifications
A propos de cet article
Pipe chase assemblies are used when service lines are too numerous to fit into a standard Atlas upright.
  • Easy to access with removable panels
  • Color: Black
Assemblies include three metal panels and brackets. Panels are removable for access.
Ordering information: Order one upright when using with existing upright.
Customers Who Bought This Also Bought