VWR® Atlas Pipe Chase Assemblies
# de catalogue VWRPC426-W
Fournisseur: VWR International
Spécifications
- Color:Snow white
- Description:Counter-Mounted Pipe Chase Assembly
- Cat. no.:VWRPC426-W
Spécifications
A propos de cet article
Pipe chase assemblies are used when service lines are too numerous to fit into a standard Atlas upright.
- Easy to access with removable panels
- Color: Black
Assemblies include three metal panels and brackets. Panels are removable for access.
Ordering information: Order one upright when using with existing upright.
Customers Who Bought This Also Bought