Value Plastics Barbed to sanitary adapter, ¹/₄" to mini sanitary, PVDF
# de catalogue MFLX31513-61
Fournisseur: Avantor Fluid Handling
Spécifications
Voir toutes les configurations de produits- Catalog No:
- MFLX31513-61
- MFLX40617-54BLK
- MFLX40617-53BLK
- MFLX31513-62
- MFLX40617-59BLK
- MFLX40617-52BLK
- MFLX40617-80BLK
- MFLX31513-66
- MFLX40617-61BLK
- Sterilisation:
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Warranty:
- Shelf life
- En cours de visualisation
Spécifications
A propos de cet article
Create a secure connection in your sanitary fitting system.
- Constructed of durable PVDF
These adapters are perfect to transition from sanitary clamp connections to a barb for flexible tubing. All adapters feature a flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow without particulate build-up. There are no sharp edges in the flow path to make cleaning very easy. These adapters offer good chemical resistance and may be sterilized by gamma irradiation, EtOH, or autoclave. Material meets the requirements for USP Class VI, ADCF, FDA, and are BPA-free.
- Barb size/tubing int.Ø (port 2):¼"
- Connection type:Sanitary to Barb
- Description:Barbed to sanitary adapter, ¹/₄" to mini sanitary, PVDF
- Sterilisation:Ethylene oxide (ETO) or gamma
- Fitting Material:PVDF
- Flow Pattern:Straight
- Sanitary Clamp Size (Port 1):¾"
Customers Who Bought This Also Bought