ContactUsLinkComponent
VWR INOCULATING LOOPS 10UL YEL ST BX1000
VWR® Inoculating Loops and Needles, Sterile
Catalog # 76544-926
Supplier:  VWR International
VWR INOCULATING LOOPS 10UL YEL ST BX1000
VWR® Inoculating Loops and Needles, Sterile
Catalog # 76544-926
Supplier:  VWR International
Specifications
  • Sterility:
    Sterile
  • Color:
    Yellow
  • Description:
    Loops
  • Size:
    10 µl
  • Cat. no.:
    76544-926
  • Packaging:
    25/pack
Specifications
About this item
High-impact polystyrene loops and needles are smooth and flexible to facilitate uniform and smooth streaking without damaging the gel surface.
  • Polystyrene fabrication
  • Smooth and flexible
  • Sterile
Needles are straight and suitable for removal of single colony specimens. Packed sterile in safe, tamperproof, zip-seal resealable bags. Disposable inoculating loops and needles do not need flaming, eliminating the risk of infection due to aerosol formation of pathogenic substances as well as cross-contamination due to improper sterilization. Loops and needles can be used under hoods without danger. Color-coded for easy size identification. Total length: 22.7 cm (87/8").
Customers Who Bought This Also Bought