Level IV Isolation Gown, Mortech
# de catalogue 76549-688
Fournisseur: Mortech Manufacturing
Spécifications
- Clothing Size:L
- W×L:25.5×48"
- Cat. no.:76549-688
- Sleeve:24 - 24.5"
Spécifications
A propos de cet article
These level 4 isolation gowns offer the highest level of protection available and are used for high risk circumstances. The lightweight, breathable design allows for all-day comfort. They offer pathogen resistance, protection against non-airborne diseases, and are an effective barrier for fluids.
- Level 4 Isolation
- 75% SMS 25% PE
- Blue
Customers Who Bought This Also Bought