ContactUsLinkComponent
CLEANER/POLISH STAINLESS STEEL
ClearSteel, Stainless Steel Cleaner and Polish Spray, Mortech
# de catalogue 76549-636
Fournisseur:  Mortech Manufacturing
CLEANER/POLISH STAINLESS STEEL
ClearSteel, Stainless Steel Cleaner and Polish Spray, Mortech
# de catalogue 76549-636
Fournisseur:  Mortech Manufacturing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    32 oz. spray bottle
  • Cat. no.:
    76549-636
Spécifications
A propos de cet article
Powerful and non-corrosive cleaner for all stainless steel surfaces. The super concentrated biodegradable formula cleans and polishes in one step.
  • Easily remove fresh or dried blood and tissue
Ideal for autopsy tables, grossing stations and all stainless steel products including instruments, lighting, coolers, body trays and carriers. Use monthly to keep your lab clean and smelling fresh
Customers Who Bought This Also Bought