Spécifications
- Size:32 oz. spray bottle
- Cat. no.:76549-636
Spécifications
A propos de cet article
Powerful and non-corrosive cleaner for all stainless steel surfaces. The super concentrated biodegradable formula cleans and polishes in one step.
- Easily remove fresh or dried blood and tissue
Ideal for autopsy tables, grossing stations and all stainless steel products including instruments, lighting, coolers, body trays and carriers. Use monthly to keep your lab clean and smelling fresh