ContactUsLinkComponent
1315-4 IV STAND 5 LEG ALUM BASE 4 HOOKS
IV Stands, Blickman
# de catalogue 76545-302
Fournisseur:  BLICKMAN INDUSTRIES
1315-4 IV STAND 5 LEG ALUM BASE 4 HOOKS
IV Stands, Blickman
# de catalogue 76545-302
Fournisseur:  BLICKMAN INDUSTRIES
Spécifications
  • Description:
    Model 1315-4 IV stand, 1" standard chrome pole on a 5 leg aluminum base, adjust height (52¹/₂ - 93¹/₂") via a Tru-Loc™ mechanism and hang IV bags on 4 hooks
  • Cat. no.:
    76545-302
  • Shipping Weight:
    9 lbs.
  • Includes:
    4-Hook removable adapter (hooks loop to the left), 1" standard chrome pole, Tru-Loc™ height adjust mechanism, height adjust: 52 ¹/₂ - 93 ¹/₂", 24" dia, 5 leg polished diecast aluminum base with Wall Saver™ tips, 2-¹/₂" black plastic hooded twin wheel casters
Spécifications
A propos de cet article
IV poles are available in high quality chrome and quality 304 stainless steel, and set on 4, 5 or 6 leg bases with casters, no projections and great balance/tip resistance.
  • Bases designed to spin 360˚ in tight spaces with great balance
  • Some bases are gravity weighted to help compensate for heavier loads
  • 2, 4, 6, or 8 (rake) hooks available for most IV pole models
  • All models come with colored secure grip tips
Our IV poles are optimal for maintaining continuous fluid delivery for every patient, available in a wide range of adjustable heights with a variety of easy locking options that can be operated with one hand.
Customers Who Bought This Also Bought