Abaque 104885 Lubricant - 1 Quart
# de catalogue MFLX75810-88
Fournisseur: NEPTUNE MIXER CO
CE Compliant
Spécifications
Voir toutes les configurations de produitsSpécifications
A propos de cet article
Replacement lubricant for the liquid end coming in four different volumnes.
- Original lubricant designed to help extend tubing life and to dissipate heat build up.
Optimize performance of your Abaque™ positive displacement pumps.
- Description:Abaque™ lubricant for positive displacement pumps, 946 ml
- Warranty:1 year
- Cat. no.:MFLX75810-88
Customers Who Bought This Also Bought