Masterflex® DB15 Male Connector; for Creating Your Own Cable
# de catalogue MFLX07595-52
Fournisseur: Avantor Fluid Handling
Spécifications
Voir toutes les configurations de produits- Catalog No:
- MFLX07595-52
- MFLX07523-94
- MFLX07595-45
- Model:
- 07595-52
- 07523-94
- 07595-45
- Control type:
- Analog Variable Speed
- Warranty:
- 2 years
- En cours de visualisation
Spécifications
A propos de cet article
Create your own cable for optimization of your Masterflex® drive.
- Choose the correct connection for your Masterflex® pump
Combine these connectors with 20-22 gauge shielded wire to create your own interface cable. Maximum cable length is 1000 ft (300 m).
- Description:DB15 male connector for creating cable
- Model:07595-52
- Cat. no.:MFLX07595-52
Customers Who Bought This Also Bought