Order Entry
ContactUsLinkComponent
MATRIX PRF 10ML FOR ORGANOID CULTURE
Corning® Matrigel® Matrix for Organoid Culture
# de catalogue CA76332-052
Fournisseur:  Corning
Numéro CAS:  
MATRIX PRF 10ML FOR ORGANOID CULTURE
Corning® Matrigel® Matrix for Organoid Culture
# de catalogue CA76332-052
Fournisseur:  Corning
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Source:
    Engelbreth-Holm-Swarm (EHS) mouse tumor
  • Description:
    Matrigel® Matrix for organoid culture
  • Size:
    10 ml
  • Storage temperature:
    –20 °C
  • Cat. no.:
    CA76332-052

Spécifications

A propos de cet article

Corning Matrigel Matrix for Organoid Culture is a solubilized basement membrane preparation that has been optimized for organoid culture.

  • Quality tested for ability to form and maintain stable 3D domes/droplets on multi-well plates
  • Elastic Modulus (matrix stiffness) measured using a rheometer
  • Protein concentrations determined by Lowry method
  • Mouse colonies routinely screened for pathogens via Mouse Antibody Production (MAP) testing
  • Extensive PCR testing is performed to screen for a number of pathogens
  • Tested and found negative for bacteria, fungi, and mycoplasma
  • Endotoxin units measured by Limulus Amoebocyte Lysate assay

Verified to support both mouse and human organoid growth and differentiation from healthy and/or diseased tissue, each lot has a specific elastic modulus value indicating the stiffness of the gel formed and is qualified to form stable 3D domes commonly used in organoid culture protocols. Available in 10 ml LDEV-free, Phenol Red-free format.

Corning Matrigel Matrix for Organoid culture is extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

Customers Who Bought This Also Bought