ContactUsLinkComponent
BTL OBL 1000CC HDPE WHT PP LNRLS CS116
Oblong Bottles, High-Density Polyethylene, Wide Mouth
Catalog # 76539-378
Supplier:  Qorpak
BTL OBL 1000CC HDPE WHT PP LNRLS CS116
Oblong Bottles, High-Density Polyethylene, Wide Mouth
Catalog # 76539-378
Supplier:  Qorpak
Specifications
  • Cap size:
    53-400
  • Cap Type:
    Unlined white PP cap
  • Capacity:
    1 L (33.8 oz.)
  • Cat. no.:
    76539-378
Specifications
About this item
Natural HDPE wide mouth Oblong bottles offer a space saving design for the workbench.
  • Bottles feature a wide mouth opening, ideal for sampling solids and powders
  • HDPE construction provides an excellent moisture barrier and reliable impact resistance
They are ideal for high-speed filling, demanding pharmaceutical specifications, and the rough handling of interstate shipping.
Ordering information: Bottles 16196-299, 16196-302, and 16125-976 are supplied pre-cleaned for volatiles. Bottles 11311-580, 11311-596, 11311-598, and 11311-600 are supplied Vacuumed and Ionized.
Customers Who Bought This Also Bought