Safety Pipette Filler, Bulb Type, Bal Supply
Catalog # 76537-054
Supplier: Tip Industries
Specifications
- Color:Blue
- Description:Pipette filler, non sterile
- Cat. no.:76537-054
Specifications
About this item
This bulb-type safety pipette filler is designed for efficient one-hand operation.
- Made from natural rubber
- Includes 3 glass valves
For use with serological pipettes of varying sizes.
Customers Who Bought This Also Bought