Order Entry
ContactUsLinkComponent
INOCULATING STREAKR LOOP M GRN 3.5MM PK5
Argos Technologies® Reusable Inoculating Streakers, Antylia Scientific
Catalog # 76534-544
Supplier:  Argos Technologies
INOCULATING STREAKR LOOP M GRN 3.5MM PK5
Argos Technologies® Reusable Inoculating Streakers, Antylia Scientific
Catalog # 76534-544
Supplier:  Argos Technologies

Specifications

  • Sterility:
    Non sterile
  • Color:
    Green
  • Description:
    Inoculating streaker loops, medium
  • Material:
    Nichrome wire and aluminum handle
  • Cat. no.:
    76534-544
  • Inner Diameter:
    3.5 mm
  • Gauge:
    26

Specifications

About this item

Nichrome wire composition cools quickly and is less expensive than platinum.

  • Select, inoculate, or transfer microbiological samples

Loops are made of high quality twisted nichrome wire and attached to a light-weight aluminum handle. Handles are insulated to protect the hand during use. Loops have been designed to slide along agar surfaces without cutting or tearing.

Loops and Handles cool rapidly after sterilization and are offered calibrated (1 and 10 µl) or non-calibrated. Non-calibrated Loops and Handles are available as small, medium or large and are color coded for easy identification.

Customers Who Bought This Also Bought