ContactUsLinkComponent
CAP REPLACEMENT POLYPROPYLENE 63MM PK12
Cole-Parmer® Essentials Replacement Caps, PP, Antylia Scientific
# de catalogue 76534-398
Fournisseur:  Argos Technologies
CAP REPLACEMENT POLYPROPYLENE 63MM PK12
Cole-Parmer® Essentials Replacement Caps, PP, Antylia Scientific
# de catalogue 76534-398
Fournisseur:  Argos Technologies
Spécifications
  • Cap Size:
    63 mm
  • Material:
    PP
  • Cap Color:
    Natural
Spécifications
A propos de cet article
Ensures a leakproof seal.
  • Autoclavable
Constructed of food-grade, biologically inert polypropylene resin, these caps are ideal replacements for lost, worn out, or broken caps.
Customers Who Bought This Also Bought