Cole-Parmer® Essentials Replacement Caps, PP, Antylia Scientific
# de catalogue 76534-398
Fournisseur: Argos Technologies
Spécifications
- Cap Size:63 mm
- Material:PP
- Cap Color:Natural
Spécifications
A propos de cet article
Ensures a leakproof seal.
- Autoclavable
Constructed of food-grade, biologically inert polypropylene resin, these caps are ideal replacements for lost, worn out, or broken caps.
Customers Who Bought This Also Bought