VISCOMETER SBS HI-VI I HELI COMPLETE
# de catalogue 76533-826
Fournisseur: IKA Works
Customers Who Bought This Also Bought
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$78.90 to $874.91
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Fournisseur: VWR International
$109.33 to $364.87
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
$635.20 to $2,541.50
Sodium hydroxide 50% (w/w) in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$279.98 to $1,119.95