Masterflex® Dispensing Tip with Luer Lock, Glass, Luer Lock by 3/16" and 1/4" ID
# de catalogue MFLX78226-60
Fournisseur: Avantor Fluid Handling
Spécifications
Voir toutes les configurations de produits- Catalog No:
- MFLX78226-60
- MFLX07595-35
- MFLX77095-03
- MFLX07571-50
- MFLX07571-55
- MFLX07571-52
- MFLX07595-40
- MFLX77095-04
- MFLX77595-35
- MFLX77120-03
- MFLX07573-02
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 2 years
- 1 year
- En cours de visualisation
Spécifications
A propos de cet article
Make the most of your Masterflex® pump with connection cables, replacement parts, and other accessories.
- Constructed of glass
- Easily attach flexible tubing to the hose barb connection
- Choose accessories and replacement parts suitable for your Masterflex® pump
Convenient accessories to make your job easier.
- Description:Glass dispensing tip with Luer lock for 3/16" and 1/4" ID tubing.
- Cat. no.:MFLX78226-60
Customers Who Bought This Also Bought