ContactUsLinkComponent
Connection Tubing FEP 1/4" UNF 1/16" ID×1/8" OD×2m, 1 Fitting
Catalogue # MFLX78167-56
Fournisseur:  KNF Neuberger Inc.
1 year warranty
undefined
Connection Tubing FEP 1/4" UNF 1/16" ID×1/8" OD×2m, 1 Fitting
Catalogue # MFLX78167-56
Fournisseur:  KNF Neuberger Inc.
Supplier Number:  166330
1 year warranty
À propos de cet article
Replacement parts keep your dosing pump running smoothly.
  • Opimize performance of your KNF Remote-Control Diaphragm Liquid Dosing Pumps
For use with KNF Remote-Control Diaphragm Liquid Dosing Pumps, pumps sold separately.
  • Description:
    Connection Tubing FEP 1/4" UNF 1/16" ID x 1/8" OD x 2m, 1 Fitting
  • Environmentally Preferable:
  • Wetted materials:
    FEP
  • Warranty:
    1 year
  • Cat. no.:
    MFLX78167-56
Frequently Bought Together