ContactUsLinkComponent
In-line Filter 70um PVDF for Dosing Pumps 78167-00 to -07
In-line Filter 70um PVDF for Dosing Pumps 78167-00 to -07
# de catalogue MFLX78167-50
Fournisseur:  KNF Neuberger Inc.
In-line Filter 70um PVDF for Dosing Pumps 78167-00 to -07
In-line Filter 70um PVDF for Dosing Pumps 78167-00 to -07
# de catalogue MFLX78167-50
Fournisseur:  KNF Neuberger Inc.
  • Wetted materials:
  • PVDF
  • FFKM
  • FEP
  • FEP
  • PVDF
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • En cours de visualisation
Spécifications
A propos de cet article
Replacement parts keep your dosing pump running smoothly.
  • Opimize performance of your KNF Remote-Control Diaphragm Liquid Dosing Pumps
For use with KNF Remote-Control Diaphragm Liquid Dosing Pumps, pumps sold separately.
  • Description:
    In-line Filter 70um PVDF for Dosing Pumps 78167-00 to -07
  • Wetted materials:
    PVDF
  • Warranty:
    1 year
  • Cat. no.:
    MFLX78167-50
Customers Who Bought This Also Bought