In-line Filter 70um PVDF for Dosing Pumps 78167-00 to -07
# de catalogue MFLX78167-50
Fournisseur: KNF Neuberger Inc.
Spécifications
Voir toutes les configurations de produits- Wetted materials:
- PVDF
- FFKM
- FEP
- FEP
- PVDF
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- En cours de visualisation
Spécifications
A propos de cet article
Replacement parts keep your dosing pump running smoothly.
- Opimize performance of your KNF Remote-Control Diaphragm Liquid Dosing Pumps
For use with KNF Remote-Control Diaphragm Liquid Dosing Pumps, pumps sold separately.
- Description:In-line Filter 70um PVDF for Dosing Pumps 78167-00 to -07
- Wetted materials:PVDF
- Warranty:1 year
- Cat. no.:MFLX78167-50
Customers Who Bought This Also Bought