ContactUsLinkComponent
Masterflex® I/P High-Performance Pump Head Moutning Screws
Masterflex® I/P High-Performance Pump Head Moutning Screws
# de catalogue MFLX77600-01
Fournisseur:  Avantor Fluid Handling
Masterflex® I/P High-Performance Pump Head Moutning Screws
Masterflex® I/P High-Performance Pump Head Moutning Screws
# de catalogue MFLX77600-01
Fournisseur:  Avantor Fluid Handling
  • Pump series:
  • I/P
  • I/P
  • I/P
  • I/P
  • I/P
  • I/P
  • I/P
  • I/P
  • Warranty:
  • 2 years
  • 2 years
  • En cours de visualisation
Spécifications
A propos de cet article
Optimize performance of your Masterflex® I/P® pump heads.
  • Choose from a wide range of replacement parts and accessories
Replacement mounting screws for the 77600-62 pump head; mounts head to I/P drive.
Replacement parts to keep your Masterflex® I/P® pump heads reliable.
  • Pump series:
    I/P
  • Description:
    I/P® High-Performance Pump Head Mounting Screws
  • Compatible with:
    77600-62
  • Cat. no.:
    MFLX77600-01
Customers Who Bought This Also Bought