Masterflex® I/P High-Performance Pump Head Moutning Screws
# de catalogue MFLX77600-01
Fournisseur: Avantor Fluid Handling
Spécifications
Voir toutes les configurations de produits- Catalog No:
- MFLX77600-01
- MFLX77600-66
- MFLX77600-67
- MFLX07019-95
- MFLX77600-03
- MFLX07019-96
- MFLX77601-95
- MFLX77601-96
- Pump series:
- I/P
- I/P
- I/P
- I/P
- I/P
- I/P
- I/P
- I/P
- Warranty:
- 2 years
- 2 years
- En cours de visualisation
Spécifications
A propos de cet article
Optimize performance of your Masterflex® I/P® pump heads.
- Choose from a wide range of replacement parts and accessories
Replacement mounting screws for the 77600-62 pump head; mounts head to I/P drive.
Replacement parts to keep your Masterflex® I/P® pump heads reliable.
- Pump series:I/P
- Description:I/P® High-Performance Pump Head Mounting Screws
- Compatible with:77600-62
- Cat. no.:MFLX77600-01
Customers Who Bought This Also Bought