Masterflex® Pump Extension Cable, RJ-12, 10-ft (3 m) Length; 1/Ea
# de catalogue MFLX77095-03
Fournisseur: Avantor Fluid Handling
Spécifications
Voir toutes les configurations de produits- Catalog No:
- MFLX77095-03
- MFLX07595-35
- MFLX78226-60
- MFLX07571-50
- MFLX07571-55
- MFLX07571-52
- MFLX07595-40
- MFLX77095-04
- MFLX77595-35
- MFLX77120-03
- MFLX07573-02
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 2 years
- 1 year
- En cours de visualisation
Spécifications
A propos de cet article
Make the most of your Masterflex® pump with connection cables, replacement parts, and other accessories.
- Increase flexibility with extended cable lengths
- Choose accessories and replacement parts suitable for your Masterflex® pump
Convenient accessories to make your job easier.
- Description:Optional extension cable, RJ12; 10 ft (3 m).
- Cat. no.:MFLX77095-03
Customers Who Bought This Also Bought