Masterflex® Dual-Drive Syringe Pump RS-232 Connector Cable
# de catalogue MFLX74901-88
Fournisseur: VWR International
Spécifications
Voir toutes les configurations de produits- Catalog No:
- MFLX74901-88
- MFLX74901-87
- Model:
- 74901-88
- 74901-87
- Outlet connection:
- RS-232 Connector Cable for 74901-80
- Daisy Chain Cable for 74901-80
- Warranty:
- 1 year
- 1 year
- En cours de visualisation
Spécifications
A propos de cet article
Daisy chain up to 100 pumps.
- Connect multiple independent syringes to operate simultaneously
- Easily connect pumps
- Choose cable based on your current pump
Optimize performance of your Masterflex® Dual-Drive Syringe Pump with these accessories.
- Description:Pumps, Syringe Pumps RS-232 Connector Cable for 74901-80 Dual Drive Syringe Pump
- Model:74901-88
- Outlet connection:RS-232 Connector Cable for 74901-80
- Warranty:1 year
- Cat. no.:MFLX74901-88
Customers Who Bought This Also Bought