Suction Line Strainer, PVC, 3/8" NPT(F), 80 Mesh (stainless steel)
# de catalogue MFLX74604-14
Fournisseur: SPECIALTY MFG CO
Spécifications
Voir toutes les configurations de produits- Catalog No:
- MFLX74604-14
- MFLX74604-22
- MFLX74604-23
- MFLX74604-20
- MFLX74604-10
- MFLX74604-12
- MFLX74604-02
- MFLX74604-04
- MFLX74604-44
- MFLX74604-52
- MFLX74604-42
- MFLX74604-43
- MFLX74604-32
- MFLX74604-34
- MFLX74604-24
- MFLX74604-30
- MFLX74605-12
- MFLX74604-53
- MFLX74604-54
- Outlet connection:
- 0.375 NPT(F)
- 0.5 NPT(F)
- 0.5 NPT(F)
- 0.5 NPT(F)
- 0.375 NPT(F)
- 0.375 NPT(F)
- 0.25 NPT(F)
- 0.25 NPT(F)
- 1 NPT(F)
- 1.5 NPT(F)
- 1 NPT(F)
- 1 NPT(F)
- 0.75 NPT(F)
- 0.75 NPT(F)
- 0.5 NPT(F)
- 0.75 NPT(F)
- 0.75 NPT(F)
- 1.5 NPT(F)
- 1.5 NPT(F)
- Wetted materials:
- PVC and Stainless Steel
- PVC and Stainless Steel
- PVC and Stainless Steel
- PVC and Stainless Steel
- PVC and Stainless Steel
- PVC and Stainless Steel
- PVC and Stainless Steel
- PVC and Stainless Steel
- PVC and Stainless Steel
- PVC and Stainless Steel
- PVC and Stainless Steel
- PVC and Stainless Steel
- PVC and Stainless Steel
- PVC and Stainless Steel
- PVC and Stainless Steel
- PVC and Stainless Steel
- Nylon and Stainless Steel
- PVC and Stainless Steel
- PVC and Stainless Steel
- Warranty:
- 1 year
- 1 year
- 6 months
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 6 months
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 6 months
- 1 year
- En cours de visualisation
Spécifications
A propos de cet article
Protect pump from sediments filtering into suction line.
- PVC (polyvinyl chloride) or Nylon base material of construction
- 20, 40, 80 mesh sizes available
- Easily removable for cleaning or replacing
These suction line pump filter strainers are ideal for keeping sediment and debris out of your pump for optimal performance. Choose from PVC (polyvinyl chloride) or nylon base material in a wide range of mesh sizes for any application.
- Description:Suction Line Strainer, PVC, 3/8" NPT(F), 80 Mesh (stainless steel)
- Wetted materials:PVC and Stainless Steel
- Outlet connection:0.375 NPT(F)
- Warranty:1 year
- Cat. no.:MFLX74604-14
- Inlet Connection:0.375 NPT(F)
Customers Who Bought This Also Bought