LMI RPM-812 Service Kit for Solenoid Diaphragm Metering Pumps, 0.42 GPH
# de catalogue MFLX74513-08
Fournisseur: EQUIP SOLUTIONS
Spécifications
Voir toutes les configurations de produits- Warranty:
- None
- None
- None
- None
- None
- None
- En cours de visualisation
Spécifications
A propos de cet article
Optimize performance of your LMI Solenoid Diaphragm Metering Pumps.
- This kit is for : 812XX, 818XX, 910NX, 910SX
- Keep your pump running smoothly with these service kits
Replace wearable parts as part of a regular maintenance program to prevent failures and downtime.
- Description:Service Kit for Solenoid Diaphragm Metering Pumps. 0.42 gal./h
- Warranty:None
- Cat. no.:MFLX74513-08
Customers Who Bought This Also Bought