Stenner Compact Peristaltic Pump Stand for Horizontal Display or Wall Mounting
# de catalogue MFLX74209-96
Fournisseur: STENNER PUMP COMPANY, INC.
Spécifications
Voir toutes les configurations de produitsSpécifications
A propos de cet article
Optmize performance of your Compact Economical Peristaltic Pump.
- Compatible with Compact Economical Peristaltic Pumps
Optional stand provides stability if dipsplaying horizontally, and can also act as a wall mounting bracket.
Choose from a wide range of replacement parts and accessories to ensure top performance of your pump. Optional stand provides stability if displaying the pump horizontally, and can also act as a wall mounting bracket.
- Color:Grey
- Description:Compact Peristaltic Pump Stand for Horizontal Display or Wall Mounting
- Material:ABS
- Warranty:None
- Compatible with:74209-00 thru -06 peristaltic pumps
- Cat. no.:MFLX74209-96
Customers Who Bought This Also Bought