ContactUsLinkComponent
Stenner Compact Peristaltic Pump Stand for Horizontal Display or Wall Mounting
Stenner Compact Peristaltic Pump Stand for Horizontal Display or Wall Mounting
# de catalogue MFLX74209-96
Stenner Compact Peristaltic Pump Stand for Horizontal Display or Wall Mounting
Stenner Compact Peristaltic Pump Stand for Horizontal Display or Wall Mounting
# de catalogue MFLX74209-96
  • Warranty:
  • None
  • None
  • None
  • En cours de visualisation
Spécifications
A propos de cet article
Optmize performance of your Compact Economical Peristaltic Pump.
  • Compatible with Compact Economical Peristaltic Pumps
Optional stand provides stability if dipsplaying horizontally, and can also act as a wall mounting bracket.
Choose from a wide range of replacement parts and accessories to ensure top performance of your pump. Optional stand provides stability if displaying the pump horizontally, and can also act as a wall mounting bracket.
  • Color:
    Grey
  • Description:
    Compact Peristaltic Pump Stand for Horizontal Display or Wall Mounting
  • Material:
    ABS
  • Warranty:
    None
  • Compatible with:
    74209-00 thru -06 peristaltic pumps
  • Cat. no.:
    MFLX74209-96
Customers Who Bought This Also Bought