ContactUsLinkComponent
Stenner Compact Peristaltic Pump Tubing Assembly, M Tube with 1/4" Ferrules; 2/Pk
Stenner Compact Peristaltic Pump Tubing Assembly, M Tube with 1/4" Ferrules; 2/Pk
# de catalogue MFLX74209-90
Stenner Compact Peristaltic Pump Tubing Assembly, M Tube with 1/4" Ferrules; 2/Pk
Stenner Compact Peristaltic Pump Tubing Assembly, M Tube with 1/4" Ferrules; 2/Pk
# de catalogue MFLX74209-90
  • Warranty:
  • None
  • None
  • None
  • En cours de visualisation
Spécifications
A propos de cet article
Optmize performance of your Compact Economical Peristaltic Pump.
  • Compatible with Compact Economical Peristaltic Pumps
Replacement Santoprene® peristaltic pump tubing assemblies with tubing ferrules. Pack of 2.
Choose from a wide range of replacement parts and accessories to ensure top performance of your pump. Optional stand provides stability if displaying the pump horizontally, and can also act as a wall mounting bracket.
  • Color:
    Opaque cream
  • Description:
    Compact Peristaltic Pump Tubing Assembly, M Tube with 1/4" Ferrules; 2/Pk
  • Material:
    Santoprene™
  • Warranty:
    None
  • Compatible with:
    74209-00
  • Cat. no.:
    MFLX74209-90
Customers Who Bought This Also Bought