Stenner Compact Peristaltic Pump Tubing Assembly, M Tube with 1/4" Ferrules; 2/Pk
# de catalogue MFLX74209-90
Fournisseur: STENNER PUMP COMPANY, INC.
Spécifications
Voir toutes les configurations de produitsSpécifications
A propos de cet article
Optmize performance of your Compact Economical Peristaltic Pump.
- Compatible with Compact Economical Peristaltic Pumps
Replacement Santoprene® peristaltic pump tubing assemblies with tubing ferrules. Pack of 2.
Choose from a wide range of replacement parts and accessories to ensure top performance of your pump. Optional stand provides stability if displaying the pump horizontally, and can also act as a wall mounting bracket.
- Color:Opaque cream
- Description:Compact Peristaltic Pump Tubing Assembly, M Tube with 1/4" Ferrules; 2/Pk
- Material:Santoprene™
- Warranty:None
- Compatible with:74209-00
- Cat. no.:MFLX74209-90
Customers Who Bought This Also Bought