ContactUsLinkComponent
Walchem Z20PC-PK Service Kit For 74126-20, -25 And 74127-20, -25
Walchem Z20PC-PK Service Kit For 74126-20, -25 And 74127-20, -25
# de catalogue MFLX74126-28
Fournisseur:  EQUIP SOLUTIONS
Walchem Z20PC-PK Service Kit For 74126-20, -25 And 74127-20, -25
Walchem Z20PC-PK Service Kit For 74126-20, -25 And 74127-20, -25
# de catalogue MFLX74126-28
Fournisseur:  EQUIP SOLUTIONS
  • Warranty:
  • 2 years
  • En cours de visualisation
Spécifications
A propos de cet article
Extend the life of your pump.
  • Easy and quick maintenance
Easy to install, these service kits avoid costly downtime of your pump.
Delivery: Includes diaphragm and retainer, valve guides, valve seats, valve balls, valve gaskets, and o-rings.
  • Description:
    Service Kit For 74126-20. -25 And 74127-20. -25
  • Warranty:
    2 years
  • Cat. no.:
    MFLX74126-28
Customers Who Bought This Also Bought