Micropump MI0043 Service Kit for 74004-03, -11; 1/Ea
# de catalogue MFLX74005-03
Fournisseur: MICROPUMP INC
Spécifications
Voir toutes les configurations de produits- Catalog No:
- MFLX74005-03
- MFLX07144-47
- MFLX07144-42
- MFLX07144-38
- MFLX07144-34
- MFLX07002-10
- MFLX74445-91
- MFLX74005-73
- MFLX07001-41
- MFLX07003-12
- MFLX07001-89
- MFLX74005-31
- MFLX07003-14
- MFLX74005-23
- MFLX74005-29
- MFLX74005-19
- MFLX74005-07
- MFLX74005-21
- MFLX74005-05
- MFLX74005-01
- MFLX74445-93
- MFLX74005-27
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- En cours de visualisation
Spécifications
A propos de cet article
Extend the life of your gear pump head with these service kits.
- Optimize performance of your Masterflex pumps
Easy to install, these service kits avoid costly downtime of your gear pump head.
- Description:Service Kit for 74004-03, -11
- Warranty:1 year
- Cat. no.:MFLX74005-03
Customers Who Bought This Also Bought