ContactUsLinkComponent
Neptune Flow checker; for models 73803-00, -30, -40; 73804-00, -30, -40
Neptune Flow checker; for models 73803-00, -30, -40; 73804-00, -30, -40
# de catalogue MFLX73805-00
Fournisseur:  NEPTUNE MIXER CO
Neptune Flow checker; for models 73803-00, -30, -40; 73804-00, -30, -40
Neptune Flow checker; for models 73803-00, -30, -40; 73804-00, -30, -40
# de catalogue MFLX73805-00
Fournisseur:  NEPTUNE MIXER CO
  • Warranty:
  • 1 year
  • En cours de visualisation
Spécifications
A propos de cet article
Easily meansure flow of your Feed Verification Diaphragm Pump.
  • Optimize performance of your Neptune Metering Pumps
These flow checkers can be mounted directly onto the discharge end of the pumps. The checker is a simple oval gear meter that measures the pump output and then transmits one pulse for every one ml of flow. It requires 4.5 to 25 VDC at a 20 mA maximum. Pump sold separately.
  • Description:
    Flow checker; for models 73803-00. -30. -40; 73804-00. -30. -40
  • Warranty:
    1 year
  • Cat. no.:
    MFLX73805-00
Customers Who Bought This Also Bought