Compression Fitting, Male Pipe Adapter (Straight), 316 SS, 1/8" OD×1/8" NPT
# de catalogue MFLX31406-34
Fournisseur: MIDWEST VALVE & FITTING INC
Spécifications
Voir toutes les configurations de produits- Sterilisation:
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Warranty:
- LTD life time
- LTD life time
- LTD life time
- LTD life time
- LTD life time
- LTD life time
- LTD life time
- En cours de visualisation
Spécifications
A propos de cet article
Excellent choice to connect rigid tubing.
- Great chemical resistance
- Easy to assemble the fitting to the tubing
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through the included sealed ferrule and using the nut to compress down to form a secure seal.
Very good chemical resistance. Pressure limitation determined by the tubing, not the fitting. Usable from -40 to 200ºF (-40 to 93ºC). Sterilize by either ethylene oxide, autoclave, or gamma irradiation.
- Connection type:Compression to Thread
- Description:Compression Fitting, Male Pipe Adapter (Straight), 316 SS, 1/8" OD×1/8" NPT
- Sterilisation:Autoclave, ethylene oxide (ETO), or gamma
- Warranty:LTD life time
- Fitting Material:Stainless Steel 316
- Flow Pattern:Straight
- Compression Size / Tube O.D. (Port 1):⅛"
- Thread Size NPT(M) (Port 2):⅛"
Customers Who Bought This Also Bought