Fitting, Stainless Steel, Compression Nut Assembly with Ferrule, 1/8"
# de catalogue MFLX31212-66
Fournisseur: MIDWEST VALVE & FITTING INC
Spécifications
Voir toutes les configurations de produits- Catalog No:
- MFLX31212-66
- MFLX31212-70
- MFLX31212-71
- MFLX31212-67
- MFLX31212-69
- MFLX31212-68
- MFLX31212-63
- MFLX31212-65
- MFLX31212-64
- Sterilisation:
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- En cours de visualisation
Spécifications
A propos de cet article
Can handle pressures up to 225 psi.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rated to 220 psi maximum at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).
- Connection type:Compression Nut Assembly with Ferrule
- Description:Fitting, Stainless Steel, Compression Nut Assembly with Ferrule, ⅛"
- Warranty:1 year
- Fitting Material:Stainless Steel 316
- Flow Pattern:Straight
- Compression Size / Tube O.D. (Port 1):⅛"
Customers Who Bought This Also Bought