ContactUsLinkComponent
Adapter for 1/4" ID Flexible Tubing, Stainless Steel
Adapter for 1/4" ID Flexible Tubing, Stainless Steel
# de catalogue MFLX07104-81
Fournisseur:  FLUID METERING INC
Adapter for 1/4" ID Flexible Tubing, Stainless Steel
Adapter for 1/4" ID Flexible Tubing, Stainless Steel
# de catalogue MFLX07104-81
Fournisseur:  FLUID METERING INC
  • Wetted materials:
  • Fluoropolymer
  • Warranty:
  • 1 year
  • 1 year
  • En cours de visualisation
Spécifications
A propos de cet article
Ideal adapters for plastic case pump heads.
  • Optimize performance of your FMI RH Series Piston Pumps
Adapters for your FMI RH series piston pump heads.
  • Description:
    Stainless Steel Adapter for Flexible Tubing, 1/4" ID
  • Warranty:
    1 year
  • Cat. no.:
    MFLX07104-81
Customers Who Bought This Also Bought