Masterflex® Pump Head to Drive Adapter Kit, Series Q
# de catalogue MFLX07104-48
Fournisseur: FLUID METERING INC
Spécifications
Voir toutes les configurations de produitsSpécifications
A propos de cet article
Easily adapt your pump head to your drive with these adapter kits.
- Choose adapter kit suitable for use with your Masterflex pump head
Use these adapter kits to attach Micropump® A-Mount pump heads to your own motor or Masterflex® L/S® pump drives.
- Description:Series Q pump head to drive adapter kit
- Warranty:1 year
- Cat. no.:MFLX07104-48
Customers Who Bought This Also Bought