ContactUsLinkComponent
Masterflex® Pump Head to Drive Adapter Kit, Series Q
Masterflex® Pump Head to Drive Adapter Kit, Series Q
# de catalogue MFLX07104-48
Fournisseur:  FLUID METERING INC
Masterflex® Pump Head to Drive Adapter Kit, Series Q
Masterflex® Pump Head to Drive Adapter Kit, Series Q
# de catalogue MFLX07104-48
Fournisseur:  FLUID METERING INC
  • Warranty:
  • 1 year
  • En cours de visualisation
Spécifications
A propos de cet article
Easily adapt your pump head to your drive with these adapter kits.
  • Choose adapter kit suitable for use with your Masterflex pump head
Use these adapter kits to attach Micropump® A-Mount pump heads to your own motor or Masterflex® L/S® pump drives.
  • Description:
    Series Q pump head to drive adapter kit
  • Warranty:
    1 year
  • Cat. no.:
    MFLX07104-48
Customers Who Bought This Also Bought