Flux Pumps Air-Powered Drum Pump Motor, 1/2 HP, 9000 rpm; 17 cfm@85 psi
# de catalogue MFLX07070-60
Fournisseur: FLUX PUMPS CORP
Spécifications
Voir toutes les configurations de produits- Catalog No:
- MFLX07070-60
- MFLX07070-70
- Air consumption:
- 28.9 m³/h (17 ft.³/min)
- Max. viscosity:
- 1200 cP
- 1200 cP
- Min. motor RPM:
- Motor hp:
- 1/2
- 1/2
- Power:
- Air
- Air
- Warranty:
- 6 months
- 6 months
- En cours de visualisation
Spécifications
A propos de cet article
Configurable to your requirements.
- High-viscosity tube set handles fluids with viscosities to 50,000 cp!
Air-operated motors are ideal for hazardous duty applications or where electricity is not available. Models 07070-60 and -70 are fitted with a muffler and include a grounding post. Model 07070-70 also has a unique trigger valve handle—press switch for intermittent pumping; lock switch for continuous use.
- Max. viscosity:1200 cP
- Description:Air-Powered Drum Pump Motor, 1/2 HP, 9000 rpm; 17 cfm@85 psi (without trigger)
- Power:Air
- Motor hp:1/2
- Air consumption:28.9 m³/h (17 ft.³/min)
- Warranty:6 months
- Cat. no.:MFLX07070-60
- Max. Motor RPM:9000 rpm
Customers Who Bought This Also Bought