Micropump 81518 Gear Pump Head Adapter Kit, 56C face drive, standard magnets
# de catalogue MFLX07003-99
Fournisseur: MICROPUMP INC
Spécifications
Voir toutes les configurations de produitsSpécifications
A propos de cet article
Easily adapt your pump head to your drive with these adapter kits.
- Choose adapter kit suitable for use with your Masterflex pump head
Use these adapter kits to attach Micropump® A-Mount pump heads to your own motor or Masterflex® L/S® pump drives.
- Description:Gear Pump Head Adapter Kit, 56C face drive, standard magnets
- Cat. no.:MFLX07003-99
Customers Who Bought This Also Bought