Micropump Economical drive for A-Mount pump heads, 150 to 4000 rpm, 0 to 24 VDC
# de catalogue MFLX07003-90
Fournisseur: MICROPUMP INC
Spécifications
Voir toutes les configurations de produits- Catalog No:
- MFLX07003-90
- MFLX07144-97
- MFLX07144-95
- Air consumption:
- 18 m³/h (11 ft.³/min)
- Duty cycle continuous:
- Yes
- Yes
- Yes
- Max. air inlet pressure:
- 5.5 bar (80 psig)
- Max. temperature:
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- Min. motor RPM:
- 300 rpm
- 500 rpm
- Min. temperature:
- 4 °C (40 °F)
- 4 °C (40 °F)
- 4 °C (40 °F)
- Motor enclosure type:
- PM brush, TENV
- TENV
- PSC
- Motor hp:
- 0.09
- 0.13 to 0.29
- 0.17
- Warranty:
- 1 year
- 1 year
- 1 year
- En cours de visualisation
Spécifications
A propos de cet article
A variety of low-cost drives for numerous applications.
- Magnetic coupling minimizes friction—all horsepower is transmitted directly to the gears
- Hundreds of hours of dependable leakproof service
These Economical Drives for Micropump A-Mount Pump Heads are designed with magnetic coupling to minimize friction. Choose from AC motor drives, DC motor drives, explosion-proof drives, or air-powered drives.
Explosion-proof and air-powered models are ideal for safe pumping in hazardous areas. Explosion-proof drives are rated for NEMA Class 1, Groups C and D locations.
- Description:Economical drive for A-Mount pump heads, 150 to 4000 rpm, 0 to 24 VDC
- Height:76 mm
- Power:0 - 24 VDC; 3 A
- Duty cycle continuous:Yes
- Min. motor RPM:300 rpm
- Motor enclosure type:PM brush, TENV
- Motor hp:0.09
- Min. temperature:4 °C (40 °F)
- Max. temperature:60 °C (140 °F)
- Warranty:1 year
- Cat. no.:MFLX07003-90
- Max. Motor RPM:4000 rpm
- Length:10.2 cm
- Width:7.6 cm
Customers Who Bought This Also Bought